UniStore™ Reagent Bottles, Wide Mouth, HDPE
Catalog #: 470349-760
Supplier: United Scientific Supplies
Specifications
- Capacity:4000 ml
- Material:HDPE
- Cat. no.:470349-760
Specifications
About this item
Bottles are for general laboratory use and for storing, shipping, and packing liquids.
- Translucent
- Pliable
UniStore™ wide mouth HDPE reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant. Excellent chemical resistance to most acids, bases, and alcohols.
Ordering information: Polypropylene caps are included.
Customers Who Bought This Also Bought