MOUNTING PLATE DSK MNT FOR 3MM DIE
Catalog #: 76636-364
Supplier: FIREBIRD OPTICS
Customers Who Bought This Also Bought
Sodium hydroxide 0.5 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
135,56 $ to 1 610,31 $
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Supplier: Ricca Chemical
175,32 $ to 1 620,42 $
Water Standards, CRM, Karl Fischer Titration, Hydranal™, Fluka™, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
469,66 $
Tryptic Soy Agar (TSA), Soybean Casein Digest Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
39,00 $