CROSS FLOW TIT SAPH 3/8 INCH
Catalog #: 76636-314
Supplier: FIREBIRD OPTICS
Customers Who Bought This Also Bought
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS, redistilled, VWR Chemicals BDH®
Supplier: VWR International
$335.08 to $1,295.24
Water, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$73.87 to $302.61
Phenol : chloroform : iso-amyl alcohol (25:24:1) in saturated aqueous solution, pH 5.2 Ready-to-Use
Supplier: Thermo Scientific Chemicals
$247.60
Anti-IgG1, k isotype ctrl Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: MOPC-21]
Supplier: Biolegend
$565.64
Pierce™ DNA Extraction Buffer, Phenol:Chloroform and Tris Buffer, Thermo Scientific
Supplier: Thermo Scientific
$628.97
Methanol-[D₄] ≥99.8% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Supplier: MilliporeSigma
$374.70
Sodium hypochlorite 5% available chlorine in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$195.79 to $1,600.59