CROSS FLOW SS 3 3/8 INCH
Catalog #: 76636-312
Supplier: FIREBIRD OPTICS
Customers Who Bought This Also Bought
Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$775.79 to $2,413.44
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$214.30 to $2,349.04
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Zinc sulfate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur. volumetric solution, Supelco®
Supplier: MilliporeSigma
$146.34 to $878.09
Potassium, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, , VWR® BDH®
Supplier: VWR International
$338.38