C OF C: GAST23.PFS.A S/N: 1169414012
Catalog #: MFLX45510-91
Supplier: MICROPUMP INC
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Dichloromethane ≥99.8% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$389.38 to $1,030.72