MOUNTING PLATE DSK FOR 8MM DIE
Catalog #: 76636-370
Supplier: FIREBIRD OPTICS
Customers Who Bought This Also Bought
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$377.44 to $992.90
Chloroform ≥99.8% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$782.44 to $2,358.46
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Supplier: Air-Tite
$73.48 to $4,408.00
Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$885.46 to $3,541.82
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15 × 100 mm plate, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
$42.99