Order Entry
ContactUsLinkComponent
HYDROGEN PEROXIDE STERILANT 950ML CS4
Bioquell Hydrogen Peroxide Sterilant Case
Catalog #: 76635-446
Supplier:  Eco Lab
CAS Number:  
HYDROGEN PEROXIDE STERILANT 950ML CS4
Bioquell Hydrogen Peroxide Sterilant Case
Catalog #: 76635-446
Supplier:  Eco Lab
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Hydrogen Peroxide Sterilant- For the Bioquell BQ50, SeQure, and ProteQ
  • Size:
    950 mL - 4 bottles per case
  • Cat. no.:
    76635-446

Specifications

About this item

Bioquell Hydrogen Peroxide Sterilant is 35% w/w Aqueous Hydrogen Peroxide Solution used specifically for Ecolab’s Bioquell bio-decontamination systems to kill microorganisms on exposed surfaces.

  • EPA registered sterilant 35% w/w aqueous hydrogen peroxide solution (EPA Registarion number 72372-1-86703)
  • Evaluated for use with Bioquell bio-decontamination hydrogen peroxide vapor systems from Ecolab
  • Designed with RFID label technology for traceability and auditing
  • Breathable foam membrane to allow venting to allow through-cap venting
  • Only one active ingredient
  • Proven material compatibility when used with Bioquell bio-decontamination systems
  • Provides efficacious validated cycles when used with Bioquell bio-decontamination systems
  • Vapor form breaks down in to water vapor and oxygen
  • 15-month shelf life while maintaining stability

Bioquell Hydrogen Peroxide Sterilant is supported with peer-reviewed scientific data supporting efficacy. The 35% aqueous hydrogen peroxide solution is designed for use with Bioquell hydrogen peroxide vapor technology. The solution has been researched, tested and evaluated to ensure it consistently delivers a 6-log sporicidal kill. Bioquell technology users can validate the process using industry standard Bioquell Geobacillus stearothermophilus biological indicators.

Certifications: EPA Registration Number 72372-1-86703
Customers Who Bought This Also Bought