Order Entry
ContactUsLinkComponent
HYQ ADCF-MAB LM 500ML
HyClone™ ADCF-mAb media, Cytiva
Catalog #: CA16777-540
Supplier:  Cytiva
CAS Number:  
HyClone™ ADCF-mAb media, Cytiva
Catalog #: CA16777-540
Supplier:  Cytiva
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ADCF-MAb™ with L-Glutamine and sodium bicarbonate, without phenol red
  • Size:
    500 ml
  • Storage temperature:
    2...8 °C
  • Media Format:
    Liquid
  • Cat. no.:
    CA16777-540

Specifications

About this item

Protein-free and animal-derived component-free medium

  • Animal-derived component-free (ADCF) formulation
  • Designed for high cell yield and monoclonal antibody (MAb) production
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities

ADCF-mAb liquid medium contains poloxamer 188 and L-glutamine, and does not contain phenol red. Liquid medium is also available without L-glutamine to support the glutamine synthetase (GS) gene expression system. ADCF‑mAb powder medium does not contain poloxamer 188, L-glutamine, phenol red, or sodium bicarbonate. ADCF-mAb is available in liquid and powder formats in user-friendly packaging.

HyClone ADCF-mAb cell culture medium is protein-free and contains no animal-derived components. This regulatory-friendly medium is developed to increase the process yields for the manufacture of antibodies and antibody fragments for therapeutic use in a variety of engineered hybridoma and myeloma cell lines.

Customers Who Bought This Also Bought