Trypsin Protease, HyClone™
Catalog #: CA82003-690
Supplier: Cytiva
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Species:Pig
- Description:Trypsin, 0.05% 1X, with 0.5 g porcine trypsin in HBSS with 0.2 g/L EDTA, without calcium, magnesium (liquid)
- Size:500 ml
- Storage conditions:Less than 0 °C
- Enzyme name:Trypsin
- Formulation:Trypsin, 0.05% 1X Solution, with 0.5 g Porcine Trypsin (1:250/L) in HBSS with 0.2 g/L EDTA, without Calcium or Magnesium
- Cat. no.:CA82003-690
Specifications
About this item
Dissociation reagents, such as trypsin, are used to detach and disaggregate adherent cells from the surface or substrate to which they are attached, as well as for dissociating tissues for primary cell culture.
- Derived from porcine pancreas
- Gamma irradiated prior to hydration and filling
- Formulated without calcium and magnesium
Customers Who Bought This Also Bought