To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Serum format:Frozen
- Sterility:Sterile filtered
- Description:Bovine calf serum, iron supplemented, defined
- Size:500 ml
- Storage temperature:Less than –10 °C
- Cat. no.:CA16777-022
Specifications
About this item
HyClone™ iron-supplemented bovine calf serum contains additional iron to boost serum transferrin levels to physiological levels.
- Sourced in the USA with complete traceability back to original source
- Cost-effective alternative to FBS
- Age of animal at time of collection approximately 16 to 24 weeks
- Low in antibodies and high in growth factors
- Virus panel testing according to 9 CFR 113.53
- Supplemented with iron and growth-promoting factors
HyClone Iron-Supplemented Bovine Calf Serum, U.S. origin is collected by venipuncture from formula-fed veal calves, which produce exceptionally high levels of transferrin. This serum is carefully supplemented with iron to load serum transferrin levels to physiological levels. Bovine calf serum products, when supplemented, can provide three to four times as much available iron as FBS. In many applications, the performance of calf serum will equal or even surpass that of FBS.
Iron-supplemented Bovine Calf Serum, U.S. origin is filtered through sequential 100 nm (0.1 µm) pore size-rated filters. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.