To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Serum format:Frozen
- Sterility:Sterile filtered
- Description:Characterized fetal bovine serum, Canadian origin, heat inactivated
- Size:500 ml
- Hemoglobin:≤25 mg/dl
- Endotoxin:≤25 EU/ml
- Storage temperature:‒10 °C
- Cat. no.:CA76236-336
Specifications
About this item
HyClone Characterized Fetal Bovine Serum, Canadian origin, uses "True Pool" processing technology to ensure consistency in the quality of serum.
- Assayed for a variety of critical components and biochemical characteristics
- Endotoxin-tested: certificate of analysis confirms ≤25 EU/ml endotoxin
- Post-filtration treatments
The final serum is assayed for provides important proteins, hormones, growth factors, metabolites, and nutrients for that support your healthy cell culture process. Assay results are included in the certificate of analysis and biochemical assay list.
Gamma-irradiated and heat-inactivated FBS options are available to further reduce the risk of adventitious agents, particularly when manufacturing products for use in humans or animals. The irradiation process is validated to deliver a minimum dose of 25 kGy.