Order Entry
ContactUsLinkComponent
MEDIA DME LOW LIQUID 1L
HyClone™ Classical Liquid Media, Dulbeccos Modified Eagles Medium, Low Glucose, HyClone products (Cytiva)
Catalog #: CA16777-127
Supplier:  Cytiva
CAS Number:  
MEDIA DME LOW LIQUID 1L
HyClone™ Classical Liquid Media, Dulbeccos Modified Eagles Medium, Low Glucose, HyClone products (Cytiva)
Catalog #: CA16777-127
Supplier:  Cytiva
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    DMEM/Low: with 1000 mg/L glucose, 4.0 mM L-glutamine, and 110 mg/L sodium pyruvate
  • Size:
    1000 ml
  • Storage temperature:
    2...8 °C
  • Cat. no.:
    CA16777-127

Specifications

About this item

DMEM Low Glucose was the original modification of Basal Medium Eagle (BME) used to grow mouse embryonic cells. DMEM, Low Glucose is suitable for the culturing of many immortalised cell lines from humans, monkeys, hamsters, rats, mice, chicken and fish. DMEM contains four times more amino acids, vitamins and other supplementary components than Basal Medium Eagle (BME), the base media from which DMEM was derived. As DMEM contains no growth-promoting agents or proteins, supplementation with FBS is recommended for optimal performance.

  • Contains 5,6 mM glucose
  • Produced with WFI quality water
  • 0.1 μm sterile filtered
  • Manufactured using ISO 9001:2000 certified processes
Customers Who Bought This Also Bought