ContactUsLinkComponent
GIBCO SOY PEPTONE 100
Soy 100, Gibco™ (a part of Thermo Fisher Scientific)
Catalog #: CA76628-930
CAS Number:  
Soy 100, Gibco™ (a part of Thermo Fisher Scientific)
Catalog #: CA76628-930
CAS Number:  
Specifications
  • Description:
    Soy 100, animal origin-free (soy)
  • Size:
    500 g
  • Cat. no.:
    CA76628-930
Specifications
About this item
Soy 100 is a highly digested soy protein peptone that is high in nucleosides and free amino acid content. It has high concentrations of vitamins and carbohydrates, which is recommended for the cultivation of a wide variety of microbial organisms as well as varied mammalian cell culture applications.
Soy 100 is also part of the Peptone Preview Pak which contains:
Cotton 100, UF - rich in nucleoside content and digested to enhance peptide content. It is processed to provide higher concentrations of vitamins, which can help maintain performance in mammalian and microbial processes.
Wheat 100, UF - ultra-filtered supplement and good source of glutamine, which makes it ideal for mammalian cell culture applications. It is digested for enhanced peptide content resulting in a low concentraion of free amino acids and carbohydrates.
Soy 100 - highly digested soy protein peptone that is high in nucleosides and free amino acid content. It has high concentrations of vitamins and carbohydrates and is recommended for a wide variety of processes.
Pea 100, UF - ultra-filtered peptone digested to enhance nucleoside and free amino acid content. The high total amino acid content and low carbohydrate content are similar to many high-performance animal origin peptones.
Yeast 100 - designed to enhance the content of vitamins and other key nutritional components found in yeast. It is intended for nutritional supplementation for mammalian, microbial, and insect cell culture.
Soy 100 can be blended with other animal origin-free peptones, such as wheat or yeast, when looking for replacements for animal origin peptones. Soy 100 is also part of the Gibco Peptone Preview Pak, which features our new wheat, cotton, soy, and yeast peptones. These peptones provide unique nutritional profiles compared to other cell culture supplements. The Preview Pak includes five 100 gram samples.
Customers Who Bought This Also Bought