Bacto™ Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Catalog #: CA76628-830
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Description:Bacto™ Peptone
- Size:500 g
- Cat. no.:CA76628-830
- Source:Bovine, Porcine
Specifications
About this item
Bacto Peptone is an enzymatic digest of bovine and porcine animal proteins. The nutritive value of Bacto Peptone is largely dependent on a combination of a negligible quantity of proteases, high concentration of complex constituents, and a unique amino acid content that makes this peptone a strong nitrogen source.
- A strong nitrogen source
- Unique amino acid content
- High concentration of complex constituents
- Vaccines for animal health applications
- Production of biotherapeutics
- Mammalian cell culture
- Microbial fermentation
Customers have successfully used this supplement with a variety of cell types including Saccharomyces cerevisiae, Escherichia coli, Pasteurella multocida, Vibrio cholerae, and porcine kidney cells to produce biotherapeutics and vaccines for animal health applications.
Certifications: LUO, MUO
Customers Who Bought This Also Bought