J.T.Baker® Robotic Tips Type HT 2
Catalog #: 49010-1115
Supplier: Ritter, part of Avantor
Specifications
- Volume:50 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- Tip Color:Black
- Packed:12× 5 Racks, 96 each, blistered
- Filter:+
- Sterile:+
Specifications
About this item
Suitable for use on third party robotic instruments Hamilton Microlab Star and Nimbus. Available in conductive version, with or without filter.
- Manufactured under cleanroom conditions (cleanroom Class 8)
- State-of-the-art quality control
- Ideal for high throughput analysis
- Barcoded trays for automated identification
- Packed in blister
J.T.Baker® robotic tips type HT 2 are suitable for third party robotic instruments Hamilton Microlab Star, Nimbus, and other robotic instruments using Hamilton’s CO-RE or CO-RE II pipetting head.
The validation of the tips on the instrument for specific applications is within the responsibility of the user.
Hamilton, Microlab Star™, Nimbus® and CO-RE® are registered trademarks of Hamilton company.
Customers Who Bought This Also Bought