Order Entry
ContactUsLinkComponent
SANITIZER HI-PH 4OZ CONC UNIT ST CS24
DECON-PHENE PLUS®, VELTEK
Catalog #: 76628-014
Supplier:  Veltek Associates
SANITIZER HI-PH 4OZ CONC UNIT ST CS24
DECON-PHENE PLUS®, VELTEK
Catalog #: 76628-014
Supplier:  Veltek Associates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    DECON-PHENE PLUS®
  • Size:
    4 oz.
  • Cat. no.:
    76628-014
  • Sterility:
    Sterile
  • Form:
    Concentrate

Specifications

About this item

DECON-PHENE Plus disinfects controlled areas such as those in compounding pharmacies, biopharmaceutical, pharmaceutical, medical device, industrial facilities, diagnostic manufacturing facilities, and research laboratories. DECON-PHENE Plus is compatible with most non-porous hard surface materials and can be used on surfaces in aseptic filling and gowning rooms, in general manufacturing areas, and in laboratories.

  • Delivered with lot specific analytical, irradiation, and sterility data, tested to current USP compendium
  • Available in our convenient, one-step, ready-to-use, SimpleMix System made with USP Water for Injection (0.25 EU/mL)
  • Multiple convenient container sizes – Unit Dose, 16 oz, 1 Gallon, and 200 L either sterile or non-sterile
Customers Who Bought This Also Bought