Order Entry
ContactUsLinkComponent
TAQ 5U/UL 250U KEY EXTRA
VWR® TAQ DNA Polymerase
Catalog #: 76620-434
Supplier:  VWR International
CAS Number:  
TAQ 5U/UL 250U KEY EXTRA
VWR® TAQ DNA Polymerase
Catalog #: 76620-434
Supplier:  VWR International
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    5 U/μl
  • Description:
    Taq DNA polymerase with 10x key buffer and 10x extra buffer
  • Size:
    250 units
  • Cat. no.:
    76620-434
  • Units:
    Each

Specifications

About this item

VWR® Taq DNA polymerase is an ultra-pure, thermostable, recombinant DNA polymerase, which provides robust PCR performance in a wide range of PCR applications, without time-consuming optimization.

  • A thermostable recombinant DNA polymerase
  • High product yield
  • dUTP incorporation possible
  • Processes up to 5 kb
  • Leaves a 3'dA overhang

Taq DNA polymerase concentration: 5 Units/μl

10X Key Buffer: Tris-HCl pH 8.5; (NH₄)₂SO₂, 15 mM MgCl₂, 1% Tween® 20
10X Extra Buffer: Tris-HCl pH 8.3; KCl, 15 mM MgCl₂, 1% Tween®20
10X Key Buffer (Mg²⁺ free): Tris-HCl pH 8.5; (NH₄)₂SO₂, 1% Tween® 20
10X Extra Buffer (Mg²⁺ free): Tris-HCl pH 8.3; KCl, 1% Tween®20

Taq DNA polymerase is isolated from Thermus aquaticus, and has a molecular weight of approximately 94 kDa. VWR® Taq DNA Polymerase has both a 5' to 3' DNA polymerase and a double strand 5' to 3' exonuclease activity. The enzyme lacks a 3'→5' exonuclease activity (no proofreading ability). It leaves an A' overhang, which makes the enzyme ideal for TA cloning.

Taq DNA polymerase is suitable for standard testing, routine PCR, screening and high-throughput testing.

Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.

Additional MgCl₂ is included with each kit to enable optimization.

Key Buffer (NH⁴⁺) gives a superior amplification signal (high yield) minimizing the need for optimization of the Mg²⁺ concentration or the annealing temperature in most primer-template systems. Extra Buffer is a traditional potassium (K⁺) buffer. Extra Buffer promotes high specificity, but careful optimization of primer annealing temperatures and Mg²⁺ concentrations may be required.

Taq DNA polymerase is also available in a glycerol-free format for automation and lyophilization.

Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought