To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:dNTP Mix 100 mM
- Size:200 μmoL
- Units:Each
- Volume:2× 1 ml
Specifications
About this item
Ready to use molecular biology grade dNTP mixes and dNTP sets.
- Available as pre-mixed 40 or 100 mM solutions , or as sets of individual 100 mM dNTP solutions
- Both pre-mixed and sets have been functionally tested in PCR
- Purity >99% by HPLC
- Supplied in solution at pH 7.3 to 7.5
The dNTP mix is designed to save hands-on time for researchers, and reduce the possibility of contamination by reducing pipetting. The dNTP mix and dNTP solutions are convenient for use in DNA polymerization reactions, DNA labelling and sequencing processes.