To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:PCR Ladder, 100 to 3000 bp
- Volume:1× 0.5 ml
- Lanes:100 lanes (5 μl)
Specifications
About this item
DNA ladders are supplied in a loading buffer that is ready to use on agarose and polyacrylamide DNA gels. The ladders are suitable with TBE, TAE, SB and LB electrophoresis systems.
- Supplied in ready to use loading buffer
- For direct loading and easy visualisation
- Clear, distinct bands
- Blue dye front running at 100 to 300 bp at 0.5 to 1.5% agarose
Mini DNA Ladder molecular range: 100 to 500 bp
Low DNA Ladder molecular range: 100 to 1000 bp
PCR Ladder molecular range: 100 to 3000 bp
Mini DNA Ladder, Low DNA Ladder and PCR Ladder are supplied in 0.5 mL packs for 100 lanes.