Order Entry
ContactUsLinkComponent
LOW DNA LADDER 100 1000 BP 100 ANES
VWR® DNA Ladders
Catalog #: 76620-564
Supplier:  VWR International
CAS Number:  
LOW DNA LADDER 100 1000 BP 100 ANES
VWR® DNA Ladders
Catalog #: 76620-564
Supplier:  VWR International
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Low DNA Ladder, 100 to 1000 bp
  • Volume:
    1× 0.5 ml
  • Lanes:
    100 lanes (5 μl)

Specifications

About this item

DNA ladders are supplied in a loading buffer that is ready to use on agarose and polyacrylamide DNA gels. The ladders are suitable with TBE, TAE, SB and LB electrophoresis systems.

  • Supplied in ready to use loading buffer
  • For direct loading and easy visualisation
  • Clear, distinct bands
  • Blue dye front running at 100 to 300 bp at 0.5 to 1.5% agarose

Mini DNA Ladder molecular range: 100 to 500 bp

Low DNA Ladder molecular range: 100 to 1000 bp

PCR Ladder molecular range: 100 to 3000 bp

Mini DNA Ladder, Low DNA Ladder and PCR Ladder are supplied in 0.5 mL packs for 100 lanes.

Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought