To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:5 U/µl
- Description:Tempase Hot Start DNA Polymerase with 10X Key Buffer and 10X Combination Buffer
- Size:500 units
- Cat. no.:76620-494
- Units:Each
Specifications
About this item
TEMPase Hot Start DNA Polymerase is a chemically modified version of VWR® Taq DNA Polymerase and is activated by 15 minutes of heat treatment.
- Reaction set-up at room temperature
- dUTP incorporation possible
- Increased sensitivity, specificity and product yield
- Detection of low abundance targets
- Processes up to 4 kb
TEMPase Hot Start DNA Polymerase concentration: 5 Units/μl
TEMPase Hot Start DNA Polymerase is suitable for detection of low abundance targets, screening, multiplexing, direct colony PCR and real-time PCR applications
TEMPase Hot Start DNA Polymerase has been designed to diminish the formation of non-specific priming events during reaction set-up and the first ramp of thermal cycling.
Chemical inactivation of TEMPase Hot Start DNA Polymerase has proven highly effective compared to other inactivation methods such as antibody inactivation. The chemically modified enzyme withstands longer periods of time at room temperature without non-specific PCR amplifications. This feature is useful if you need pre-incubation at elevated temperatures, for example in case of UNG treatment at 50 °C prior to PCR.
TEMPase Hot Start DNA polymerase is also available in a glycerol-free format for automation and lyophilization.
Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.
TEMPase Hot Start DNA Polymerase is usually supplied in kits with Key Buffer and Combination Buffer. 10X Key Buffer promotes high specificity over a broad range of annealing temperatures and Mg2+ concentrations. The Key Buffer also works well when dealing with difficult templates, e.g. GC-rich DNA sequences. 10X Combination Buffer is a balanced ammonium-potassium PCR buffer. Combination Buffer results in tolerance towards optimization of primer annealing temperatures and Mg2+ concentrations. Additional MgCl₂ is included with each kit to enable optimization.