Smiths Medical® Fitting, Polycarbonate, Three-Way, Large-Bore Stopcock with Male Luer Lock; 1000/PK
Catalog #: MFLX30600-23BLK
Supplier: SMITHS MEDICAL ASD INC
Specifications
- Description:Fitting, Polycarbonate, Three-Way, Large-Bore Stopcock with Male Luer Lock; 1000/PK
- Wetted materials:Polycarbonate
- Warranty:Out of box
- Cat. no.:MFLX30600-23BLK
- Flow Pattern:Three-Way
- Inlet Port:Female luer
- Outlet Port:Male luer lock
Specifications
About this item
Enhance fluid flow with a larger bore than standard luer stopcocks.
- Three-way flow pattern
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Large-bore adapters are perfect for handling higher flow volumes in process while using connections similar to luer systems.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
- Description:Fitting, Polycarbonate, Three-Way, Large-Bore Stopcock with Male Luer Lock; 1000/PK
- Wetted materials:Polycarbonate
- Warranty:Out of box
- Cat. no.:MFLX30600-23BLK
- Flow Pattern:Three-Way
- Inlet Port:Female luer
- Outlet Port:Male luer lock
Customers Who Bought This Also Bought