ContactUsLinkComponent
INFUSION SET WINGED 23G 3/4INX12IN BX50
Sol-Care® Safety Winged Infusion Sets, Sol M
Catalog #: 76582-718
Sol-Care® Safety Winged Infusion Sets, Sol M
Catalog #: 76582-718
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Length:
    ³/₄"
  • Cat. no.:
    76582-718
  • Tubing Length:
    12"
  • Gauge:
    23
Specifications
About this item
Sol-Care safety winged infusion sets are intended for short-term IV medication administration.
  • Thin-walled needle minimizes patient discomfort
  • Flexible, color-coded wings
  • DEHP-free, kink-resistant tubing
  • Compatible with luer connectors
Each infusion set is equipped with a sliding safety mechanism that is activated after the infusion has completed, helping to prevent needlestick injuries.
Customers Who Bought This Also Bought