VWR® Premium Thickness Nitrile Gloves
Catalog # 76582-344
Supplier: VWR International
Specifications
- Glove Color:Blue
- Glove size:S
- Ambidextrous/Hand-specific:Ambidextrous
- Disposable / reusable:Disposable
- Latex/Latex-free:Latex-free
- Powdered/Powder-free:Powder-free
- Length:9.5" (24.13 cm)
- ASTM No.:ASTM No. D6319
- Cuff style:Beaded
- Finger thickness:6 mil
- Palm thickness:4 mil
- Cuff thickness:3 mil
- Tensile strength:Min 14 (before and after aging)
- Ultimate elongation:Min 500 (before aging), Min 400 (after aging)
- Cat. no.:76582-344
Specifications
About this item
Ambidextrous 6 mil gloves are extra-soft and comfortable for everyday use. They protect hands from a broad range of chemicals.
- 6 mil finger thickness
- Nonsterile
- Latex-free
- Extra-soft and comfortable
- Powder-free
- Beaded cuff allows easy donning
Ideal for use in industrial and laboratory applications. Made of premium compounded nitrile for high manufacturing quality measures and product consistency. Gloves protect against biohazards and a broad range of chemicals. Composition is 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins. Finger textured gloves feature rolled, beaded cuffs.
Certifications: Gloves meet and exceed ASTM 6319 specification for Nitrile Examination Gloves for Medical Application.
Packaging: Gloves are packaged 100 per box, 10 boxes per case.
Frequently Bought Together