ContactUsLinkComponent
BIOHAZARD BAGS AUTOCLAVABLE CLEAR CS200
Biohazard Bags, Autoclavable, Mopec
Catalog #: 76579-390
BIOHAZARD BAGS AUTOCLAVABLE CLEAR CS200
Biohazard Bags, Autoclavable, Mopec
Catalog #: 76579-390
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Color:
    Clear
  • Description:
    Autoclavable biohazard bags
  • Cat. no.:
    76579-390
Specifications
About this item
Mopec’s autoclavable biohazard bags are specifically designed with seamless bottoms for durability and leak-resistance, and are made from a heavy-duty, puncture-resistant polypropylene. These bags can also withstand temperatures up to 250 ˚F during autoclaving. Each bag features the OSHA-standard 'Biohazard' symbol and words 'Hazardous Waste'.
  • Dimensions: 26L×12.2"W
  • Made from a heavy-duty, puncture-resistant polypropylene
  • Can withstand temperatures up to 250 ˚F during autoclaving
  • Feature OSHA-standard 'Biohazard' symbol and words 'Hazardous Waste'
  • Feature large, white marking panel for use with permanent markers
  • Seamless bottoms for durability and leak-resistance
Sold by the case of 200 bags.
Customers Who Bought This Also Bought