SureSTART™ Electronic Hand-Held Crimpers and Decrimpers, Level 2 High Throughput Applications, Thermo Scientific
Catalog #: TS60180-EDCB11
Supplier: Thermo Scientific
Specifications
- Description:Electronic hand held battery operated decrimper, suitable for 11 mm crimp caps. Generation 4
- Cat. no.:TS60180-EDCB11
Specifications
About this item
Use Thermo Scientific™ electronic hand-held vial crimpers and decrimpers to attach or remove crimp caps from vials, respectively.
These hand-held, high-power crimpers and decrimpers are available in 8, 11, 13, and 20 mm sizes and allow you to seal or remove crimp caps with a simple, one-handed push of a button. Use for crimping of vials with aluminum, pressure-release, and bi-metal caps. The electronic crimper's message window conveniently provides all-important information at one glance. This device is performance level 3, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses.
Customers Who Bought This Also Bought