FLOW RATE HYDRAULIC FLOW METER SAE 50GPM
Catalog #: MFLX00343-IL
Supplier: BADGER METER INC
Customers Who Bought This Also Bought
Color Standard, APHA / HzAen (Platinum-Cobalt), Color = 500, Ricca Chemical Company
Supplier: Ricca Chemical
$1,429.06
Accessories for Single-Sample Osmometer, Model 3250, Advanced Instruments
Supplier: Advanced Instruments
$554.99
Hydrochloric acid 36.5 - 38.0%, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$103.36 to $1,240.08
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Supplier: Ricca Chemical
$160.67 to $1,597.95
Copper ≥99.9% (metals basis), shot 1-10 mm (0.04-0.4 in)
Supplier: Thermo Scientific Chemicals
$106.00
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Supplier: MilliporeSigma
$156.54 to $571.97