Order Entry
ContactUsLinkComponent
SPOT PLATE-12 WELLS PORCELAIN
Eisco Porcelain Spot Plates
Catalog #: 470345-848
Supplier:  Wards
SPOT PLATE-12 WELLS PORCELAIN
Eisco Porcelain Spot Plates
Catalog #: 470345-848
Supplier:  Wards

Specifications

  • Well Ø:
    22 mm
  • Description:
    Porcelain spot plate with 12 depressions
  • L×W:
    4.5×3.75"
  • Cat. no.:
    470345-848

Specifications

About this item

Spot plates made of high quality glazed porcelain which can be cleaned and reused.

  • Spot plate made of high quality porcelain glazed on the top and sides
  • Wells measure 22 mm in diameter
  • Can be cleaned and reused
  • Perfect for using with liquid or small solid samples

Plate is glazed on top and sides and can be washed and reused. Useful for comparing results among students in a lab, or in classroom demonstrations.

Customers Who Bought This Also Bought