Order Entry
ContactUsLinkComponent
1000 ML MEASURING JUG
Eisco Plastic Measuring Jugs, Short Form
Catalog #: 470344-656
Supplier:  Wards
1000 ML MEASURING JUG
Eisco Plastic Measuring Jugs, Short Form
Catalog #: 470344-656
Supplier:  Wards

Specifications

  • Capacity:
    1000 ml
  • Description:
    Plastic measuring jug, short form
  • Cat. no.:
    470344-656

Specifications

About this item

Translucent polypropylene measuring jug with short form (Euro design), raised graduations, tapered spout, and handle.

  • Made of premium quality polypropylene plastic
  • Superior chemical durability and heat tolerance (can withstand temperatures up to 135 °C)
  • Autoclave, microwave and dishwasher safe.
  • Excellent resistance to strong acids, alcohols and bases
  • Measuring jug is translucent and features excellent optical clarity with raised graduations and specifications
  • Tapered spout for easy pouring and transferring of liquids
  • Equipped with a handle for easy and convenient lifting

Jug features excellent optical clarity and easy to read, raised graduations and specifications. 50 ml subdivisions. Features a spout for easy pouring, and handle for stable handling and convenient lifting. Microwave and dishwasher safe.

Customers Who Bought This Also Bought