ContactUsLinkComponent
MICRO GLASS DISTILLATION UNIT
Micro Glass Distillation Kit
Catalog #: 470344-628
Supplier:  Wards
MICRO GLASS DISTILLATION UNIT
Micro Glass Distillation Kit
Catalog #: 470344-628
Supplier:  Wards
Specifications
  • Description:
    Micro glass distillation kit
  • Cat. no.:
    470344-628
Specifications
About this item
Designed for micro-scale distillation, this 9-piece kit includes all the necessary components with 19/26 joints, enabling precise and efficient distillation processes in a compact and convenient setup for laboratory use.
  • Distillation is the process of separating the component or substances from a liquid mixture by selective evaporation and condensation
  • Kit includes 9 pieces : 1 - 50 ml conical flask, 1 - round bottom boiling flask 25 ml, 1 - liebig condenser 100 mm, 1 - still head, 1 - receiver adapter, 1 - funnel 35 mm, 1 - 50 ml beaker, 1 - 10 ml burette ( class A 0.1 ml graduations, ±0.02 tolerance), and 1 - spirit burner
  • Some glass pieces are threaded to make a tight connection with plastic connectors to make a tight fit during operation
  • All pieces are made of high quality borosilicate glass 3.3
  • Clamps, stand, or thermometer are not included with this kit
Eisco Borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.
Customers Who Bought This Also Bought