Order Entry
ContactUsLinkComponent
BRUSH TT-NYLON L100MM PK12
Eisco Nylon Test Tube Brushes with Fan-Shaped Ends
Catalog #: 470344-330
Supplier:  Wards
BRUSH TT-NYLON L100MM PK12
Eisco Nylon Test Tube Brushes with Fan-Shaped Ends
Catalog #: 470344-330
Supplier:  Wards

Specifications

  • Bristle Material:
    Nylon
  • Brush Head Diameter:
    35 mm
  • Handle Material:
    Stainless steel
  • Description:
    Nylon test tube brush
  • Total length:
    280 mm
  • Cat. no.:
    470344-330

Specifications

About this item

Gentle nylon test tube brushes.

  • Designed to scrub narrow glassware
  • Fanned end that allows you to easily clean rounded surfaces and corners
  • Moderate flexibility allows easy modification to handle shape

These semi-stiff nylon bristle brushes are perfect for scrubbing narrow tube-shaped glassware such as test tubes and culture tubes. Each brush features a fanned end that allows you to easily clean rounded surfaces and corners. Each brush has a stainless steel, twisted handle that is semi-flexible, allowing you to modify the shape as needed. Each handle has a looped end, for easy hanging.

Customers Who Bought This Also Bought