VWR® Atlas Pipe Chase Assemblies
Catalog #: VWRPC606-W
Supplier: VWR International
Specifications
- Color:Snow white
- Description:Counter-Mounted Pipe Chase Assembly
- Cat. no.:VWRPC606-W
Specifications
About this item
Pipe chase assemblies are used when service lines are too numerous to fit into a standard Atlas upright.
- Easy to access with removable panels
- Color: Black
Assemblies include three metal panels and brackets. Panels are removable for access.
Ordering information: Order one upright when using with existing upright.
Customers Who Bought This Also Bought