Order Entry
ContactUsLinkComponent
SW VWR LAB TABLE NO UPRIGHTS 36HX36DX72L
VWR® Lab Tables
Catalog #: WB363672-GK
Supplier:  VWR International
SW VWR LAB TABLE NO UPRIGHTS 36HX36DX72L
VWR® Lab Tables
Catalog #: WB363672-GK
Supplier:  VWR International

Specifications

  • Base/Leg Material:
    Powder coated steel
  • Description:
    VWR® Lab table frame, no uprights
  • W×D×H:
    72×36×36"
  • Capacity:
    1000 lbs. (SEFA)
  • Base/Leg Color/Finish:
    Snow white

Specifications

About this item

A four-leg, flexible lab table system designed for any application.

  • Powder coated steel
  • Chemical resistant epoxy resin
  • Optional shelving
  • Optional casters
  • Accepts suspended cabinets
  • Accepts fixtures (with uprights)
  • 1000 lbs. capacity

This table system includes a number of accessories that can be added very easily, creating your ideal workstation.

Certifications: SEFA tested (SEFA 3 and 10).
Ordering information: Work tops and table frames are sold separately. Tables shipped knocked down, and will need assembly. Professional Assembly is highly recommended.
Customers Who Bought This Also Bought