Viral inactivation solution, Ultrapure Bioreagent, J.T.Baker®
Catalog #: CAJTX191-01
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Plastic bottle
- Environmentally Preferable:Safer Human and Environmental Health
- Size:10 ml
- Environmental certification:EPA safer choice chemical
- Cat. no.:CAJTX191-01
Specifications
Specification Test Results
- Identification (by FTIR):Passes Test
- Appearance:Passes Test
- Purity by HPLC:min 95%
- Assay:Passes Test
- Acid value:max 10
- Viscosity (25°C) :For information only
- Density:0.9 – 1.1 g/mL
- Limit of free ethylene oxide (Target <1 ppm):For information only
- Total microbial count (No growth detected):Passes Test
- Endotoxin:For information only
- Protein reactive impurities (100x Diluted):For information only
- Arsenic (As):For information only
- Cadmium (Cd):For information only
- Lead (Pb):For information only
- Mercury (Hg):For information only
- Cobalt (Co):For information only
- Nickel (Ni):For information only
- Vanadium (V):For information only
- Silver (Ag):For information only
- Gold (Au):For information only
- Iridium (Ir):For information only
- Osmium (Os):For information only
- Palladium (Pd):For information only
- Platinum (Pt):For information only
- Rhodium (Rh):For information only
- Ruthenium (Ru):For information only
- Selenium (Se):For information only
- Thallium (Tl):For information only
- Barium (Ba):For information only
- Chromium (Cr):For information only
- Copper (Cu):For information only
- Lithium (Li):For information only
- Molybdenum (Mo):For information only
- Antimony (Sb):For information only
- Tin (Sn):For information only
- J.T.Baker®, Viral inactivation solution, Ultrapure Bioreagent:
Specification Test Results
About this item
:Viral inactivation solution
Density:0.9 – 1.1 g/ml
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
Storage Temperature:Ambient
Customers Who Bought This Also Bought