Order Entry
ContactUsLinkComponent
SOLUTION 301 CELL LYSIS 10ML
J.T.Baker® Cell Lysis Solution, Ultrapure Bioreagent
Catalog #: CAJTX193-01
   Safer Human and Environmental Health
CAS Number:  
SOLUTION 301 CELL LYSIS 10ML
J.T.Baker® Cell Lysis Solution, Ultrapure Bioreagent
Catalog #: CAJTX193-01
   Safer Human and Environmental Health
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Packaging:
    Plastic bottle
  • Description:
    Cell Lysis Solution, Ultrapure Bioreagent
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Size:
    10 ml
  • Environmental certification:
    EPA safer choice chemical
  • Cat. no.:
    CAJTX193-01

Specifications

About this item

J.T. Baker® Cell Lysis Solution. Optimized multi-component, ready-to-use solution for extraction and recovery of AAV particles from HEK293 or SF9 cells. The product offers quantitative recovery through efficient cell lysis and protection of vector particles against shear stress. The solution is formulated with non-ionic detergents which are biodegradable and octyl phenol ethoxylate free.

  • Comparable cell lysis performance to octyl phenol ethoxylate
  • Improved protection of viral particles
  • Reduced foaming and viscosity
  • 0.2 µm filtered solution

Proprietary formulation. Not hazardous according to regulation 29 CFR 1910.1200 (OSHA HCS)

Chemical Specifications of Non-GMP Cell lysis solution:
Refractive Index: 1.1-1.5
Appearance: Passes Test
pH: For information only
Viscosity (25°C): For information only
Density: 0.9 – 1.2 g/mL
Total microbial count: No growth detected
UV-Vis (max Absorption) at 380-450 nm: For information only

Customers Who Bought This Also Bought