PLgel Individual Pore Size, Agilent Technologies
Catalog #: CAAGPL11108520
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Column Format:Analytical
- I.D.:7.50 mm
- Length:600 mm
- Max. pressure:150 bar
- Particle size:5 µm
- Pore size:100 Å
- pH range:1-14
- Temperature limit:150 °C
- Cat. no.:CAAGPL11108520
- Packing Material:Fully Porous
- Phase:Plgel
- Exclusion Limit:0 - 5000 Da
Specifications
About this item
PLgel individual pore size columns offer high resolution over a mol wt range in organic solvents.
- PLgel individual pore size GPC columns offer the highest resolution over a narrow molecular weight range
- May be stacked to extend the molecular weight range
- Recommended for very small and very large polymers, which can be difficult to resolve using more common PLgel MIXED columns
PLgel individual pore size GPC columns offer the highest resolution within a narrow molecular weight range. Columns may be stacked to extend the molecular weight range. Individual pore columns are generally recommended for very small and very large polymers, which can be difficult to resolve using more common PLgel MIXED columns.
Customers Who Bought This Also Bought