CrossLab Capillaries for Waters LC Systems, Agilent Technologies
Catalog #: CAAG8005-0826
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- I.D.:0.02"
- Length:10'
- Description:Agilent tubing, ¹/₁₆" Ext.Ø, similar to Waters WAT026804
- Material:Stainless steel
- Cat. no.:CAAG8005-0826
Specifications
About this item
Stainless steel capillaries and tubing for selected Waters HPLC systems.
- Made of ultra-clean stainless steel tubing
- Ideal for most standard applications, and the best choice for reliable high-pressure sealing
- Smooth inner surface for the lowest backpressure
- Precision cutting for burr-free, square-cut ends without inner-diameter distortion
- Both ends accept standard fittings
- Predefined lengths for specific path locations to avoid peak broadening
- Individually tested to ensure reproducibility
- Available in pre-swaged
Agilent offers a range of capillaries and tubing made from stainless steel for selected Waters HPLC systems. Used with the right fittings, they are ideal for LC system plumbing, tight, leak-free connections, and zero dead volumes.
Customers Who Bought This Also Bought