CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Stainless Steel screens, 0.75 inch Ext. Ø for 100 ml tubes, 20-mesh, 864 µm, for BIO-DIS, Pack of 25
Catalog # CAAG27-2310
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Supplier #: 27-2310
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Catalog # CAAG27-2310
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Jump to:
Product Details & Documents
Several reciprocating vessel screens are available for the BIO-DIS reciprocating cylinder apparatus.
- Reciprocating cylinder screens for BIO-DIS in stainless steel and polypropylene are available in different mesh sizes
Several reciprocating vessel screens are available for the Agilent BIO-DIS reciprocating cylinder apparatus, which is ideal for automatic dissolution testing of dosage forms that require different types of media.
Specifications
- Description:Stainless Steel screens, 0.75 inch Ext. Ø for 100 ml tubes, 20-mesh, 864 µm, for BIO-DIS, Pack of 25