CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Catalog #: CAAG27-2110
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Stainless Steel screens, 1.25 inch Ext. Ø for 300 ml tubes, 100-mesh, 140 µm, for BIO-DIS, Pack of 25
Specifications
About this item
Several reciprocating vessel screens are available for the BIO-DIS reciprocating cylinder apparatus.
- Reciprocating cylinder screens for BIO-DIS in stainless steel and polypropylene are available in different mesh sizes
Several reciprocating vessel screens are available for the Agilent BIO-DIS reciprocating cylinder apparatus, which is ideal for automatic dissolution testing of dosage forms that require different types of media.
Customers Who Bought This Also Bought