Dissolution Evaporation Covers, Agilent Technologies
Catalog #: CAAG27-1500
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Evaporation cover, USP, for 300 ml tube, for BIO-DIS
Specifications
About this item
A variety of evaporation covers are available to fit your apparatus and vessels.
- The evaporation covers automatically travel with the apparatus driveunit using either the alignment posts or DDM assemblies
- The standard evaporation cover on the 708-DS is designed to allow less than 1% evaporative loss over a 24-hour period under normal operating conditions
- Agilent also offers other low loss covers that can reduce evaporative loss even further
A variety of evaporation covers are available to fit your apparatus and vessels. Evaporation plugs are used to minimize evaporation by sealing unused ports in the evaporation covers.
Customers Who Bought This Also Bought