VWR® Inoculating Loops and Needles, Sterile
Catalog #: 76544-936
Supplier: VWR International
Specifications
- Sterility:Sterile
- Color:Blue
- Description:Loops
- Size:1 µl
- Packaging:25/pack
Specifications
About this item
High-impact polystyrene loops and needles are smooth and flexible to facilitate uniform and smooth streaking without damaging the gel surface.
- Polystyrene fabrication
- Smooth and flexible
- Sterile
Needles are straight and suitable for removal of single colony specimens. Packed sterile in safe, tamperproof, zip-seal resealable bags. Disposable inoculating loops and needles do not need flaming, eliminating the risk of infection due to aerosol formation of pathogenic substances as well as cross-contamination due to improper sterilization. Loops and needles can be used under hoods without danger. Color-coded for easy size identification. Total length: 22.7 cm (87/8").
Customers Who Bought This Also Bought