ContactUsLinkComponent
BLADEFLASK BLADE REMOVER RED
Qlicksmart® Blade Remover, BladeFLASK, Mortech
Catalog #: 76549-464
BLADEFLASK BLADE REMOVER RED
Qlicksmart® Blade Remover, BladeFLASK, Mortech
Catalog #: 76549-464
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Qlicksmart® blade remover, BladeFLASK
  • Cat. no.:
    76549-464
Specifications
About this item
The Qlicksmart® Blade Removal System is a medical safety device specially designed, engineered and manufactured to comply with recently revised OSHA Bloodborne Pathogens Standards. Qlicksmart® offers single-handed removal and immediate containment of contaminated scalpel blades which dramatically reduces the risk and cost of injury to the user.
  • Holds 100 blades
  • Fits blade nos. 10, 10a, 11, 12, 15, 20, 21, 22, 23, 24
  • Fits handle nos. 3 (AJ140), 4 (AJ101), 4L (AJ102), 5, 7, 9
  • Easy mounting with optional AJ151 mounting bracket
  • OSHA compliant
Features a built-in counter and automatic shut off when full.
BodyTextAccessories: The mounting bracket is necessary for the Qlicksmart® Blade Removal System to comply with recently revised OSHA Bloodborne Pathogens Standards. Qlicksmart® offers single-handed removal and immediate containment of contaminated scalpel blades which dramatically reduces the risk and cost of injury to the user when properly mounted.
Customers Who Bought This Also Bought