Rapid Response™ COVID-19 Antigen Rapid Test Cassettes - At Home, BTNX
Catalog # CA76540-464
Supplier: BTNX INC CA
CAS Number:
Specifications
- Description:SARS-CoV-2 antigen
- Size:2 packs
- Cat. no.:CA76540-464
Specifications
About this item
The Rapid Response® COVID-19 antigen rapid test cassette - at home is an in vitro immunoassay which detects the nucleocapsid proteins of SARS-CoV-2, the virus that causes COVID-19. The test uses nasal swabs taken from asymptomatic or symptomatic people within 7 days of symptom onset.
- Authorized for self-testing
- Results in just 10 minutes
- Compact kit sizes
- Uses nasal swab samples
- Easy-to-follow instructions with illustrations
The Rapid Response® COVID-19 antigen rapid test cassette - at home is authorized for home use in a non-laboratory setting with direct anterior nasal (nares) swab samples for: unobserved self-collection for individuals aged 18 years or older, adult-supervised self-collection for individuals age 14 or older, and adults collecting from children two years or older.
This test is authorized under the Health Canada Interim Order (IO #342928).
Frequently Bought Together