Order Entry
ContactUsLinkComponent
Hose Fittings, Band Clamps, for 6" OD Hose, 25/Pk
Hose Fittings, Band Clamps, for 6" OD Hose, 25/Pk
Catalog #: MFLX33350-18
Supplier:  VWR International
Hose Fittings, Band Clamps, for 6" OD Hose, 25/Pk
Hose Fittings, Band Clamps, for 6" OD Hose, 25/Pk
Catalog #: MFLX33350-18
Supplier:  VWR International
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • Currently Viewing

Specifications

About this item

Quick and easy assembly with band clamp tool.

  • Constructed of durable stainless steel

Band Clamps are ideal for tightening hose over barbed fittings. Constructed of stainless steel. All band clamps require a band clamp tool to install.

  • Tubing O.D.:
    152.4 mm (6")
  • Description:
    Hose Fittings, Band Clamps, for 6" OD Hose, 25/Pk
  • Warranty:
    1 year
  • Cat. no.:
    MFLX33350-18
Customers Who Bought This Also Bought