Masterflex® Quattroflow QF1200S Mutiple-Use Pump Chamber; 316L machined
Catalog #: MFLX76000-08
Supplier: VWR International
Specifications
See all product configurationsSpecifications
About this item
Keep your Quattroflow™ pump system up and running with these quality replacement pump heads.
- No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
- Single-use pump heads are constructed with a disposable polypropylene chamber
- Reusable pump heads are constructed of durable 316L stainless steel
Keep your Quattroflow pump system up and running with these quality replacement pump heads. The quaternary diaphragm design enables risk-free dry-running, self-priming, and a high turn-down ratio. These pump heads can safely and reliably pump plasma products, proteins, monoclonal antibodies, vaccines, and more.
Order the Masterflex® Single-use Quattroflow™ Pump Systems separately.
- Description:Mutiple-Use Pump Chamber; 316L machined stainless steel multiuse Quattroflow pump systems
- Warranty:1 year
- Cat. no.:MFLX76000-08
Customers Who Bought This Also Bought