Abaque 104885 Lubricant - 1 Quart
Catalog #: MFLX75810-88
Supplier: NEPTUNE MIXER CO
CE Compliant
Specifications
See all product configurationsSpecifications
About this item
Replacement lubricant for the liquid end coming in four different volumnes.
- Original lubricant designed to help extend tubing life and to dissipate heat build up.
Optimize performance of your Abaque™ positive displacement pumps.
- Description:Abaque™ lubricant for positive displacement pumps, 946 ml
- Warranty:1 year
- Cat. no.:MFLX75810-88
Customers Who Bought This Also Bought