Micropump Gear Pump Accessory Service Kit for 74445-05; 1/Ea
Catalog #: MFLX74445-91
Supplier: MICROPUMP INC
Specifications
See all product configurations- Catalog No:
- MFLX74445-91
- MFLX07144-47
- MFLX07144-42
- MFLX07144-38
- MFLX07144-34
- MFLX07002-10
- MFLX74005-73
- MFLX07001-41
- MFLX07003-12
- MFLX07001-89
- MFLX74005-31
- MFLX07003-14
- MFLX74005-23
- MFLX74005-29
- MFLX74005-19
- MFLX74005-07
- MFLX74005-21
- MFLX74005-03
- MFLX74005-05
- MFLX74005-01
- MFLX74445-93
- MFLX74005-27
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- Currently Viewing
Specifications
About this item
Extend the life of your gear pump head with these service kits.
- Optimize performance of your Masterflex pumps
Service kit includes: gears, bushings, and seals.
Easy to install, these service kits avoid costly downtime of your gear pump head.
- Description:Gear Pump Service Kit, for 74445-05
- Warranty:1 year
- Cat. no.:MFLX74445-91
Customers Who Bought This Also Bought