SafeMask® SofSkin® Procedure Earloop Face Masks, Medicom
Catalog #: CA76544-824
Supplier: Medicom
Specifications
- Sterility:Non sterile
- Color:Lavender
- Description:SafeMask® SofSkin® procedure earloop face masks
- Cat. no.:CA76544-824
Specifications
About this item
The highest level of protection in the softest mask.
- Non-woven spunbond outer layer provides maximum protection against fluid penetration
- Shingle pleat design prevents fluid pooling and cross-contamination
- High-quality media ensures optimal filtration and breathability
- Adjustable, enclosed nosepiece forms a strong seal to provide customized protection and comfort
- Latex-free
Made especially for sensitive skin. Lightweight, breathable inner layer softens with wear and remains cool on skin. Unique wicking technology keeps face dry and comfortable. Soft, strong, ultrasonically sealed earloops attached to outside of mask prevent pulling and irritation, and allow easy removal with less risk of self-contamination.
Procedure masks are intended to be worn by personnel during medical procedures to protect both the patient and the operating personnel from transfer of microorganisms, body fluids, and particulate material transfer.
Packaging: Masks are packaged 50/box, 10 boxes/case.
Customers Who Bought This Also Bought