Universal Precautions Lab Coats, Halyard
Catalog #: CA76540-372
Supplier: O&M HALYARD CANADA, INC CA
Specifications
- Clothing Size:Small
- Color:Blue
- Cat. no.:CA76540-372
Specifications
About this item
Halyard Health Lab Coats all feature a protective 3-layer fabric that offers exceptional resistance to blood strike-through at various levels of exposure.
- Traditional collar
- Knit cuffs
- Six snap front closure
- Three reinforced pockets
- Size/name label
Halyard Health Lab Coats all feature a back vent for added comfort during extended use, and a neck loop for hanging.
Customers Who Bought This Also Bought