BASIC PLUS* Lab Coats, Halyard
Catalog #: CA76540-350
Supplier: O&M HALYARD CANADA, INC CA
Specifications
- Clothing Size:XXL
- Color:White
- Cat. no.:CA76540-350
Specifications
About this item
HALYARD* BASIC PLUS* Lab Coats all feature a protective 3-layer fabric that offers exceptional resistance to blood strike-through at various levels of exposure.
- Knit collar and cuffs
- Five snap front closure
- Three reinforced pockets
- Neck loop for hanging
- Size/name label
Customers Who Bought This Also Bought